Learn More
Biomatik Corporation Rat Interleukin 18 Binding Protein (IL18BP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05260
Encompass_Preferred
Rat Interleukin 18 Binding Protein (IL18BP) ELISA Kit for detecting IL18BP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.251ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JLN2. Gene ID: 84388. Target Synonyms: IL18BPa; MC51L-53L-54L Homolog Gene Product; Tadekinig-alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2966