Learn More
Biomatik Corporation Rat Phospholipase A2 Group VII (LpPLA2) ELISA Kit
Supplier:  Biomatik Corporation EKU09031
Encompass_Preferred
Rat Phospholipase A2 Group VII (LpPLA2) ELISA Kit for detecting LpPLA2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.5-800pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5M7T7. Gene ID: 301265. Target Synonyms: PLA2G7; PAF-AH; PAFAH; Lp-PLA2; LDL-PLA2; Platelet Activating Factor Acetylhydrolase,Plasma; Phospholipase A2,Group VII; LDL-associated phospholipase A2; Phospholipase A2, Lipoprotein Associated. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6568