Learn More
Biomatik Corporation Mouse Immunoglobulin G2a (IgG2a) ELISA Kit
Supplier:  Biomatik Corporation EKU09371
Encompass_Preferred
Mouse Immunoglobulin G2a (IgG2a) ELISA Kit for detecting IgG2a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.075ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6611