Learn More
Biomatik Corporation Pig Interleukin 2 (IL2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05273
Encompass_Preferred
Pig Interleukin 2 (IL2) ELISA Kit for detecting IL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26891. Gene ID: 396868. Target Synonyms: TCGF; Lymphokine; T-Cell Growth Factor; Aldesleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2975