Learn More
Biomatik Corporation Rat Interleukin 2 (IL2) ELISA Kit, 96 tests
Catalog No. 501502976
Encompass_Preferred
Rat Interleukin 2 (IL2) ELISA Kit for detecting IL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17108. Gene ID: 116562. Target Synonyms: TCGF; Lymphokine; T-Cell Growth Factor; Aldesleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2976 Supplier Biomatik Corporation Supplier No. EKU05274
Rat Interleukin 2 (IL2) ELISA Kit for detecting IL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17108. Gene ID: 116562. Target Synonyms: TCGF; Lymphokine; T-Cell Growth Factor; Aldesleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.