Learn More
Biomatik Corporation Rabbit Interleukin 2 (IL2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05275
Encompass_Preferred
Rabbit Interleukin 2 (IL2) ELISA Kit for detecting IL2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O77620. Gene ID: 100302458. Target Synonyms: TCGF; Lymphokine; T-Cell Growth Factor; Aldesleukin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2977