Learn More
Biomatik Corporation Pig Corticotropin Releasing Factor (CRF) ELISA Kit
Supplier:  Biomatik Corporation EKU10750
Encompass_Preferred
Pig Corticotropin Releasing Factor (CRF) ELISA Kit for detecting CRF in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.09pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P06296. Gene ID: 100127468. Target Synonyms: CRH; Corticotropin Releasing Hormone; Corticoliberin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6755