Learn More
Biomatik Corporation Human Fucosyltransferase 2 (FUT2) ELISA Kit
Supplier:  Biomatik Corporation EKU10848
Encompass_Preferred
Human Fucosyltransferase 2 (FUT2) ELISA Kit for detecting FUT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q10981. Gene ID: 2524. Target Synonyms: SE; Se2; Sej; Secretor Status Included; Galactoside 2-Alpha-L-Fucosyltransferase 2; Secretor blood group alpha-2-fucosyltransferase; Secretor factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6782