Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 1 (FABP1) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10925
Encompass_Preferred
Human Fatty Acid Binding Protein 1 (FABP1) High-sensitive ELISA Kit for detecting FABP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.107ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07148. Gene ID: 2168. Target Synonyms: FABP-1; FABPL; L-FABP; LFABP; Liver-type fatty acid-binding protein; Fatty Acid Binding Protein 1, Liver. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6803