Learn More
Biomatik Corporation Mouse Interleukin 23 (IL23) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05296
Encompass_Preferred
Mouse Interleukin 23 Subunit Alpha (IL23a) ELISA Kit for detecting IL23a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQ14. Gene ID: 83430. Target Synonyms: P19; IL23P19; SGRF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2983