Learn More
Biomatik Corporation Mouse Interleukin 24 (IL24) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05301
Encompass_Preferred
Mouse Interleukin 24 (IL24) ELISA Kit for detecting IL24 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q925S4. Gene ID: 93672. Target Synonyms: C49A; FISP; IL10B; MDA7; Mob-5; ST16; Mda-7; Melanoma Differentiation Association Protein 7; Suppression Of Tumorigenicity 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2987