Learn More
Biomatik Corporation Human Interleukin 25 (IL25) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05303
Encompass_Preferred
The Human Interleukin 25 (IL25) ELISA Kit allows for quantitative determination interleukin 25 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9H293. Cross Activity: No significant cross-reactivity or interference between Interleukin 25 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2988