Learn More
Biomatik Corporation Mouse Interleukin 25 (IL25) ELISA Kit, 96 tests
Catalog No. 501502989
Encompass_Preferred
Mouse Interleukin 25 (IL25) ELISA Kit for detecting IL25 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VHH8. Gene ID: 140806. Target Synonyms: IL17E; IL17-E; Interleukin-17E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2989 Supplier Biomatik Corporation Supplier No. EKU05304
Mouse Interleukin 25 (IL25) ELISA Kit for detecting IL25 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VHH8. Gene ID: 140806. Target Synonyms: IL17E; IL17-E; Interleukin-17E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.