Learn More
Biomatik Corporation Rat Interleukin 25 (IL25) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05305
Encompass_Preferred
Rat Interleukin 25 (IL25) ELISA Kit for detecting IL25 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZLB1. Gene ID: 501996. Target Synonyms: IL17E; IL17-E; Interleukin-17E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2990