Learn More
Biomatik Corporation Human Reticulocalbin 3 (RCN3) ELISA Kit
Supplier:  Biomatik Corporation EKU11400
Encompass_Preferred
Human Reticulocalbin 3 (RCN3) ELISA Kit for detecting RCN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96D15. Gene ID: 57333. Target Synonyms: RLP49; EF-Hand Calcium Binding Domain; EF-hand calcium-binding protein RLP49. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6912