Learn More
Biomatik Corporation Mouse Interleukin 31 (IL31) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05319
Encompass_Preferred
Mouse Interleukin 31 (IL31) ELISA Kit for detecting IL31 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 2.36pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6EAL8. Gene ID: 76399. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2996