Learn More
Biomatik Corporation Human Interleukin 32 (IL32) ELISA Kit, 96 tests
Catalog No. 501502997
Encompass_Preferred
The Human Interleukin 32 (IL32) ELISA Kit allows for quantitative determination interleukin 32 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24001. Cross Activity: No significant cross-reactivity or interference between Interleukin 32 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2997 Supplier Biomatik Corporation Supplier No. EKU05320
The Human Interleukin 32 (IL32) ELISA Kit allows for quantitative determination interleukin 32 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24001. Cross Activity: No significant cross-reactivity or interference between Interleukin 32 and analogues was observed. Restriction: For Research Use Only.