Learn More
Biomatik Corporation Heparan Sulfate (HS) ELISA Kit
Supplier:  Biomatik Corporation EKU11875
Encompass_Preferred
Heparan Sulfate (HS) ELISA Kit for detecting HS in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 246.9-20,000pg/ml. Sensitivity: 89.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7013