Learn More
Biomatik Corporation Mouse Interleukin 33 (IL33) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05322
Encompass_Preferred
Mouse Interleukin 33 (IL33) ELISA Kit for detecting IL33 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BVZ5. Gene ID: 77125. Target Synonyms: DV27; C9ORF26; IL1F11; NFHEV; Interleukin-1 Family, Member 11; Nuclear factor from high endothelial venules. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2999