Learn More
Biomatik Corporation Rat Interleukin 34 (IL34) ELISA Kit, 96 tests
Catalog No. 501503002
Encompass_Preferred
Rat Interleukin 34 (IL34) ELISA Kit for detecting IL34 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4KM46. Gene ID: 498951. Target Synonyms: C16orf77. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3002 Supplier Biomatik Corporation Supplier No. EKU05326
Rat Interleukin 34 (IL34) ELISA Kit for detecting IL34 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4KM46. Gene ID: 498951. Target Synonyms: C16orf77. Restrictions: For Research Use Only. Not for use in diagnostic procedures.