Learn More
Biomatik Corporation Human Leucine Rich Repeat Containing G Protein Coupled Receptor 5 (LGR5) ELISA Kit
Supplier:  Biomatik Corporation EKU12237
Encompass_Preferred
Human Leucine Rich Repeat Containing G Protein Coupled Receptor 5 (LGR5) ELISA Kit for detecting LGR5 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75473. Gene ID: 8549. Target Synonyms: FEX; GPR49; GPR67; HG38; G Protein-Coupled Receptor 49. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7097