Learn More
Biomatik Corporation Mouse Interleukin 35 (IL35) ELISA Kit, 96 tests
Catalog No. 501503004
Encompass_Preferred
Mouse Interleukin 35 (IL35) ELISA Kit for detecting IL35 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43431. Gene ID: 16159. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3004 Supplier Biomatik Corporation Supplier No. EKU05328
Mouse Interleukin 35 (IL35) ELISA Kit for detecting IL35 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43431. Gene ID: 16159. Restrictions: For Research Use Only. Not for use in diagnostic procedures.