Learn More
Biomatik Corporation Human Ependymin Related Protein 1 (EPDR1) ELISA Kit
Supplier:  Biomatik Corporation EKU12260
Encompass_Preferred
Human Ependymin Related Protein 1 (EPDR1) ELISA Kit for detecting EPDR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UM22. Gene ID: 54749. Target Synonyms: MERP1; UCC1; EPDR; Upregulated In Colorectal Cancer Gene-1; Mammalian ependymin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7106