Learn More
Biomatik Corporation Dog Interleukin 4 (IL4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05330
Encompass_Preferred
The Dog Interleukin 4 (IL4) ELISA Kit allows for quantitative determination interleukin 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O77762. Cross Activity: No significant cross-reactivity or interference between Interleukin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3005