Learn More
Biomatik Corporation Rat Complement Factor I (CFI) ELISA Kit
Supplier:  Biomatik Corporation EKU12330
Encompass_Preferred
Rat Complement Factor I (CFI) ELISA Kit for detecting CFI in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.239ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUW3. Gene ID: 79126. Target Synonyms: CF-I; C3b-INA; FI; IF; KAF; Factor I; Konglutinogen-Activating Factor; C3b-Inactivator; C3B/C4B inactivator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7119