Learn More
Biomatik Corporation Pig Interleukin 4 (IL4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05337
Encompass_Preferred
Pig Interleukin 4 (IL4) ELISA Kit for detecting IL4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q04745. Gene ID: 397225. Target Synonyms: BSF1; BCGF1 ;B Cell Stimulatory Factor 1; Lymphocyte stimulatory factor 1; B Cell Growth Factor; Binetrakin; Pitrakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3010