Learn More
Biomatik Corporation Human transferrin(TF) ELISA Kit
Supplier:  Biomatik Corporation EKC31338
Encompass_Preferred
Sample Type: serum, plasma, urine, cell culture supernates, cerebrospinal fluid (CSF). Species Reactivity: Homo sapiens (Human). Detection Range: 0.023 nmol/L-1.5 nmol/L. Sensitivity: 0.006 nmol/L. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P02787. Target Synonyms: DKFZp781D0156, PRO1557, PRO2086. Restriction: For Research Use Only.
Catalog No. 50-228-7176