Learn More
Biomatik Corporation Human Interleukin 4 Receptor (IL4R) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05342
Encompass_Preferred
The Human Interleukin 4 Receptor (IL4R) ELISA Kit allows for quantitative determination interleukin 4 Receptor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24394. Cross Activity: No significant cross-reactivity or interference between Interleukin 4 Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3014