Learn More
Biomatik Corporation Mouse Interleukin 5 (IL5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05349
Encompass_Preferred
Mouse Interleukin 5 (IL5) ELISA Kit for detecting IL5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04401. Gene ID: 16191. Target Synonyms: E-CSF; ESGF; EDF; TRF; Colony-Stimulating Factor,Eosinophil; T-Cell replacing Factor; B Cell Differentiation Factor I; Eosinophil Differentiation Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3017