Learn More
Biomatik Corporation Chicken Epidermal Growth Factor, Egf Elisa Kit
Supplier:  Biomatik Corporation EKC31919
Encompass_Preferred
Chicken Epidermal Growth Factor, Egf Elisa Kit for detecting Epidermal Growth Factor, Egf in serum, plasma, tissue homogenates. Species Reactivity: Chicken (Gallus). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7238