Learn More
Biomatik Corporation Human Interleukin 5 Receptor Alpha (IL5Ra) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05352
Encompass_Preferred
The Human Interleukin 5 Receptor Alpha (IL5Ra) ELISA Kit allows for quantitative determination interleukin 5 Receptor Alpha in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78.12-5000 pg/mL. Sensitivity: 24.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q01344. Cross Activity: No significant cross-reactivity or interference between Interleukin 5 Receptor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3019