Learn More
Biomatik Corporation Chicken Immunoglobulin A,IgA ELISA Kit
Supplier:  Biomatik Corporation EKC31939
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Gallus gallus (Chicken). Detection Range: 781.25 ng/mL-50000 ng/mL. Sensitivity: 390 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7241