Learn More
Biomatik Corporation Dog Interferon Γ , Ifn-Γ Elisa Kit
Supplier:  Biomatik Corporation EKC32063
Encompass_Preferred
Dog Interferon Γ , Ifn-Γ Elisa Kit for detecting Interferon, Gamma in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P42161; IFNG. Target Synonyms: IFNGInterferon gamma; IFN-gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7252