Learn More
Biomatik Corporation Dog Myeloperoxidase (Mpo) Elisa Kit
Supplier:  Biomatik Corporation EKC32093
Encompass_Preferred
Dog Myeloperoxidase (Mpo) Elisa Kit for detecting Myeloperoxidase, Mpo in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.25-2000ng/ml. Sensitivity: 7.81ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: F1PQ52. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7255