Learn More
Biomatik Corporation Fish Adrenocorticotropic Hormone (Acth) Elisa Kit
Supplier:  Biomatik Corporation EKC32140
Encompass_Preferred
Fish Adrenocorticotropic Hormone (Acth) Elisa Kit for detecting Adrenocorticotropic Hormone (Acth) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 75-1200pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7259