Learn More
Biomatik Corporation Fish Cholecystokinin (Cck) Elisa Kit
Supplier:  Biomatik Corporation EKC32146
Encompass_Preferred
Fish Cholecystokinin (Cck) Elisa Kit for detecting Cholecystokinin(Cck) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 200-5000pg/ml. Sensitivity: 125pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: D7S9W7; CCK-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7261