Learn More
Biomatik Corporation Fish Glutathione Peroxidase (Gsh-Px) Elisa Kit
Supplier:  Biomatik Corporation EKC32150
Encompass_Preferred
Fish Glutathione Peroxidase (Gsh-Px) Elisa Kit for detecting Glutathione Peroxidase (Gsh-Px) in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 375-6000uU/ml. Sensitivity: 187.5uU/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7263