Learn More
Biomatik Corporation Rabbit Interleukin 6 (IL6) ELISA Kit, 96 tests
Catalog No. 501503029
Encompass_Preferred
Rabbit Interleukin 6 (IL6) ELISA Kit for detecting IL6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.24pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9MZR1. Gene ID: 100008733. Target Synonyms: MGI2-A; MGI2A; HGF; BSF2; HSF; IFNB2; B-Cell Stimulatory Factor-2; Hybridoma/Plasmacytoma Growth Factor; Hepatocyte Stimulating Factor; Cytotoxic T-Cell Differentiation Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3029 Supplier Biomatik Corporation Supplier No. EKU05364
Rabbit Interleukin 6 (IL6) ELISA Kit for detecting IL6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.24pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9MZR1. Gene ID: 100008733. Target Synonyms: MGI2-A; MGI2A; HGF; BSF2; HSF; IFNB2; B-Cell Stimulatory Factor-2; Hybridoma/Plasmacytoma Growth Factor; Hepatocyte Stimulating Factor; Cytotoxic T-Cell Differentiation Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.