Learn More
Biomatik Corporation Human Interleukin 6 Receptor (IL6R) ELISA Kit, 96 tests
Catalog No. 501503031
Encompass_Preferred
The Human Interleukin 6 Receptor (IL6R) ELISA Kit allows for quantitative determination interleukin 6 Receptor in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.88-120 ng/mL. Sensitivity: 0.67 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08887. Cross Activity: No significant cross-reactivity or interference between Interleukin 6 Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3031 Supplier Biomatik Corporation Supplier No. EKU05366
The Human Interleukin 6 Receptor (IL6R) ELISA Kit allows for quantitative determination interleukin 6 Receptor in Serum, Plasma, Urine, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.88-120 ng/mL. Sensitivity: 0.67 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08887. Cross Activity: No significant cross-reactivity or interference between Interleukin 6 Receptor and analogues was observed. Restriction: For Research Use Only.