Learn More
Biomatik Corporation Fish Interferon Γ , Ifn-Γ Elisa Kit
Supplier:  Biomatik Corporation EKC32159
Encompass_Preferred
Fish Interferon Γ , Ifn-Γ Elisa Kit for detecting Interferon Γ , Ifn in serum, plasma, tissue homogenates. Species Reactivity: Fish (Vertebrata). Detection Range: 20-4000pg/ml. Sensitivity: 10pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Target Synonyms: IFN-gamma,interferon gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7270