Learn More
Biomatik Corporation Fish Interleukin-8,IL-8 ELISA Kit
Supplier:  Biomatik Corporation EKC32162
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Fish. Detection Range: 31.25 pg/mL-500 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6F4N2. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7273