Learn More
Biomatik Corporation Goat Interleukin 1Β (Il-1Β) Elisa Kit
Supplier:  Biomatik Corporation EKC32210
Encompass_Preferred
Goat Interleukin 1Β (Il-1Β) Elisa Kit for detecting Interleukin 1Β(Il-1Β) in serum, plasma, tissue homogenates. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P79162; IL1B. Target Synonyms: IL1BInterleukin-1 beta; IL-1 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7282