Learn More
Biomatik Corporation Gallus Interleukin 8 (IL8) ELISA Kit, 96 tests
Catalog No. 501503036
Encompass_Preferred
The Gallus Interleukin 8 (IL8) ELISA Kit allows for quantitative determination interleukin 8 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08317. Cross Activity: No significant cross-reactivity or interference between Interleukin 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3036 Supplier Biomatik Corporation Supplier No. EKU05377
The Gallus Interleukin 8 (IL8) ELISA Kit allows for quantitative determination interleukin 8 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08317. Cross Activity: No significant cross-reactivity or interference between Interleukin 8 and analogues was observed. Restriction: For Research Use Only.