Learn More
Biomatik Corporation Horse Fibrinogen(Fbg)ELISA Kit
Supplier:  Biomatik Corporation EKC32323
Encompass_Preferred
Sample Type: serum, plasma, urine. Species Reactivity: Equus caballus (Horse). Detection Range: 23.4 ng/mL-1500 ng/mL. Sensitivity: 23.4 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7292