Learn More
Biomatik Corporation Human 3-Nitrotyrosine (3-Nt) Elisa Kit
Supplier:  Biomatik Corporation EKC32402
Encompass_Preferred
Human 3-Nitrotyrosine (3-Nt) Elisa Kit for detecting 3-Nitrotyrosine (3-Nt) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7294