Learn More
Biomatik Corporation Rabbit Interleukin 8 (IL8) ELISA Kit, 96 tests
Catalog No. 501503039
Encompass_Preferred
Rabbit Interleukin 8 (IL8) ELISA Kit for detecting IL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.16pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19874. Gene ID: 100009129. Target Synonyms: CXCL8; AMCF-I; GCP1; K60; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP1; SCYB8; TSG1; B-ENAP; Neutrophil-Activating Protein 1; Granulocyte Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3039 Supplier Biomatik Corporation Supplier No. EKU05381
Rabbit Interleukin 8 (IL8) ELISA Kit for detecting IL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.16pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19874. Gene ID: 100009129. Target Synonyms: CXCL8; AMCF-I; GCP1; K60; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP1; SCYB8; TSG1; B-ENAP; Neutrophil-Activating Protein 1; Granulocyte Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.