Learn More
Biomatik Corporation Human 5-Hydroxytryptamine, 5Ht Elisa Kit
Supplier:  Biomatik Corporation EKC32412
Encompass_Preferred
Human 5-Hydroxytryptamine, 5Ht Elisa Kit for detecting 5-Hydroxytryptamine, 5Ht in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4-800ng/ml. Sensitivity: 2ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7296