Learn More
Biomatik Corporation Human 8-iso prostaglandin F2α,8-iso-PGF2a ELISA Kit
Supplier:  Biomatik Corporation EKC32423
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 125 pg/mL-5000 pg/mL. Sensitivity: 125 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: 8-iso prostaglandin F2α,8-iso prostaglandin F2 alpha,8-iso-PGF2α,8-iso-PGF2 alpha. Restriction: For Research Use Only.
Catalog No. 50-228-7299