Learn More
Biomatik Corporation Rat Interleukin 8 Receptor Alpha (IL8Ra) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05385
Encompass_Preferred
Rat Interleukin 8 Receptor Alpha (IL8Ra) ELISA Kit for detecting IL8Ra in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70612. Gene ID: 54258. Target Synonyms: CD181; CDw128a; CXCR1; CKR1; CMKAR1; IL8R1; IL8RBA; IL8-RA; Chemokine C-X-C motif Receptor 1; High Affinity Interleukin-8 Receptor A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3041