Learn More
Biomatik Corporation Human Acetylcholinesterase, Ache Elisa Kit
Supplier:  Biomatik Corporation EKC32455
Encompass_Preferred
Human Acetylcholinesterase, Ache Elisa Kit for detecting Acetylcholinesterase (Yt Blood Group) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.94-60mU/ml. Sensitivity: 0.23mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P22303; ACHE. Target Synonyms: ACEE; ACES_HUMAN; Acetylcholinesterase; AChE; Apoptosis related acetylcholinesterase; ARACHE; N ACHE; N-ACHE; YT; YT blood group. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7301